9-(3-Bromophenyl)-10-phenylanthracene 99%
Supplier: Ambeed
|
Warning
|
A557603-1G
A557603-250MG
A557603-5G
77157-214EA
27
USD
77157-214
77157-216
77157-218
9-(3-Bromophenyl)-10-phenylanthracene 99%
9-(3-Bromophenyl)-10-phenylanthracene
Precaución: for Research Use Only
Formula:
C₂₆H₁₇Br |
MDL Number:
MFCD28100940 CAS Number: 1023674-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...