D-(-)-Citrulline 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Cit-OH

A556213-1G A556213-5G
77157-210EA 70.04 USD
77157-210 77157-212
D-(-)-Citrulline 98%
D-(-)-Citrulline

Precaución: for Research Use Only

Formula: C₆H₁₃N₃O₃
MW: 175.19 g/mol
MDL Number: MFCD00216469
CAS Number: 13594-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR