6-Methyl-1,3-benzoxazole-2-thiol 98%

Supplier: Ambeed
Warning

A556062-1G A556062-250MG A556062-5G
77157-202EA 16.48 USD
77157-202 77157-204 77157-206 77157-208
6-Methyl-1,3-benzoxazole-2-thiol 98%
6-Methyl-1,3-benzoxazole-2-thiol

Precaución: for Research Use Only

Formula: C₈H₇NOS
MDL Number: MFCD00087075
CAS Number: 23417-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR