(2S,4R)-tert-Butyl 4-fluoro-2-(hydroxymethyl)pyrrolidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A555985-1G A555985-5G
77157-200EA 161.71 USD
77157-200 77551-676
(2S,4R)-tert-Butyl 4-fluoro-2-(hydroxymethyl)pyrrolidine-1-carboxylate 98%
(2S,4R)-tert-Butyl 4-fluoro-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈FNO₃
MW: 219.25 g/mol
MDL Number: MFCD22573266
CAS Number: 1138324-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR