Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetate Hydrobromide 97%

Supplier: Ambeed
Warning

A555683-1G A555683-250MG A555683-5G A555683-100G
77157-194EA 16 USD
77157-194 77157-196 77157-198 77901-677
Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetate Hydrobromide 97%
Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetate Hydrobromide

Precaución: for Research Use Only

Formula: C₂₆H₅₁BrN₄O₆
MDL Number: MFCD11506707
CAS Number: 149353-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR