(1R,3S)-(+)-Camphoric acid 98%

Supplier: Ambeed
Warning

Synonyms: D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B

A555427-10G A555427-25G
77157-190EA 8.24 USD
77157-190 77157-192
(1R,3S)-(+)-Camphoric acid 98%
(1R,3S)-(+)-Camphoric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
MDL Number: MFCD00001375
CAS Number: 124-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR