z-Tyr-OMe 97%

Supplier: Ambeed
Warning

Synonyms: Z-Tyr-OMe, N-Benzyloxycarbonyl-L-tyrosine methyl ester

A552338-100G A552338-10G A552338-25G A552338-5G
77157-166EA 129.78 USD
77157-166 77157-168 77157-170 77157-172
z-Tyr-OMe 97%
z-Tyr-OMe

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₅
MW: 329.352 g/mol
MDL Number: MFCD00038784
CAS Number: 13512-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR