Retinol all trans 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
all-trans-Retinol, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol, Vitamin A alcohol
A550285-100MG
A550285-1G
77157-150EA
60
USD
77157-150
77157-152
Retinol all trans 98%
Retinol all trans
Precaución: for Research Use Only
Formula:
C₂₀H₃₀O |
MDL Number:
MFCD00001552 CAS Number: 68-26-8 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...