2-(4-Fluorobenzoyl)pyridine 95%

Supplier: Ambeed
Warning

A549177-1G A549177-250MG
77157-146EA 80 USD
77157-146 77157-148
2-(4-Fluorobenzoyl)pyridine 95%
2-(4-Fluorobenzoyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₈FNO
MDL Number: MFCD07699233
CAS Number: 169955-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR