1-(4-Bromophenyl)cyclopentanecarboxylic acid 98%

Supplier: Ambeed
Warning

A545825-100MG A545825-250MG A545825-5G
77157-112EA 10 USD
77157-112 77157-114 77157-116
1-(4-Bromophenyl)cyclopentanecarboxylic acid 98%
1-(4-Bromophenyl)cyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrO₂
MDL Number: MFCD00455267
CAS Number: 143328-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR