4-Butylthiobenzamide 95%

Supplier: Ambeed
Warning

A542706-100MG A542706-1G A542706-250MG A542706-5G
77157-106EA 15 USD
77157-106 77157-108 77157-110 77686-398
4-Butylthiobenzamide 95%
4-Butylthiobenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅NS
MDL Number: MFCD09025670
CAS Number: 1208077-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR