(2R,3R,4R,5R,6R)-2-((((2R,3R,4R)-3,4-Dihydroxy-4-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-6-(3,4-dihydroxyphenethoxy)-5-hydroxy-4-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl (E)-3-(3,4-dihydroxyphenyl)acrylate 95%

Supplier: Ambeed
Warning

Synonyms: Forsythoside B

A542297-10MG A542297-1MG A542297-5MG
77157-094EA 88 USD
77157-094 77157-096 77157-098
(2R,3R,4R,5R,6R)-2-((((2R,3R,4R)-3,4-Dihydroxy-4-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-6-(3,4-dihydroxyphenethoxy)-5-hydroxy-4-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl (E)-3-(3,4-dihydroxyphenyl)acrylate 95%
(2R,3R,4R,5R,6R)-2-((((2R,3R,4R)-3,4-Dihydroxy-4-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-6-(3,4-dihydroxyphenethoxy)-5-hydroxy-4-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl (E)-3-(3,4-dihydrox

Precaución: for Research Use Only

Formula: C₃₄H₄₄O₁₉
MDL Number: MFCD03424245
CAS Number: 81525-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR