4,4'-Dibromo-1,1'-binaphthalene 97%

Supplier: Ambeed
Warning

A540693-10G A540693-1G A540693-5G
77157-084EA 737.48 USD
77157-084 77157-086 77157-088
4,4'-Dibromo-1,1'-binaphthalene 97%
4,4'-Dibromo-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₂₀H₁₂Br₂
MW: 412.11 g/mol
MDL Number: MFCD00021409
CAS Number: 49610-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR