1-(2-(Aminomethyl)morpholino)ethanone 95%

Supplier: Ambeed
Danger

A540244-250MG A540244-100MG
77157-082EA 1386 USD
77157-082 77901-401
1-(2-(Aminomethyl)morpholino)ethanone 95%
1-(2-(Aminomethyl)morpholino)ethanone

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O₂
MDL Number: MFCD09892296
CAS Number: 182067-97-6
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR