(2'Z,3'E)-6-Bromoindirubin-3'-oxime 98%

Supplier: Ambeed
Danger

Synonyms: GSK 3 Inhibitor IX, BIO, 6BIO, 6-Bromoindirubin-3'-oxime, (2Z,3E)-6'-Bromo-3-(hydroxyimino)-[2,3'-biindolinylidene]-2'-one

A538818-10MG A538818-1MG A538818-50MG A538818-5MG
77157-070EA 25.75 USD
77157-070 77157-072 77157-074 77157-076
(2'Z,3'E)-6-Bromoindirubin-3'-oxime 98%
(2'Z,3'E)-6-Bromoindirubin-3'-oxime

Precaución: for Research Use Only

Formula: C₁₆H₁₀BrN₃O₂
MW: 356.18 g/mol
MDL Number: MFCD08705318
CAS Number: 667463-62-9
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR