3,4-Difluorobenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 3,4-Difluorobenzene-1-sulphonyl chloride

A537981-10G A537981-5G
77157-052EA 18.54 USD
77157-052 77157-054
3,4-Difluorobenzenesulfonyl chloride 97%
3,4-Difluorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
MDL Number: MFCD00153120
CAS Number: 145758-05-0
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR