1-Benzyloxy-4-nitrobenzene 98%

Supplier: Ambeed
Warning

A536893-100G A536893-25G A536893-5G
77157-042EA 52 USD
77157-042 77157-044 77157-046
1-Benzyloxy-4-nitrobenzene 98%
1-Benzyloxy-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₃
MDL Number: MFCD00024672
CAS Number: 1145-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR