1,2,3-Cyclohexanetriol (cis and trans mixture) 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,3-Cyclohexanetriol, Hexahydropyrogallol, Pyrogallitol

A534092-1G A534092-5G A534092-25G
77157-008EA 31 USD
77157-008 77686-377 77893-090
1,2,3-Cyclohexanetriol (cis and trans mixture) 98%
1,2,3-Cyclohexanetriol (cis and trans mixture)

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MDL Number: MFCD00060111
CAS Number: 6286-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR