JW55 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(((4-(4-Methoxyphenyl)tetrahydro-2H-pyran-4-yl)methyl)carbamoyl)phenyl)furan-2-carboxamide, N-[4-[[[[Tetrahydro-4-(4-methoxyphenyl)-2H-pyran-4-yl]methyl]amino]carbonyl]phenyl]-2-furancarboxamide

A533890-10MG A533890-1MG A533890-50MG
77157-002EA 45 USD
77157-002 77157-004 77157-006
JW55 98%
JW55

Precaución: for Research Use Only

Formula: C₂₅H₂₆N₂O₅
MDL Number: MFCD03863265
CAS Number: 664993-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR