2-Methoxy-4-(trifluoromethyl)benzyl alcohol 95%

Supplier: Ambeed
Warning

A529644-5G A529644-1G A529644-250MG
77156-994EA 140 USD
77156-994 77901-685 77901-112
2-Methoxy-4-(trifluoromethyl)benzyl alcohol 95%
2-Methoxy-4-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₉F₃O₂
MDL Number: MFCD04115979
CAS Number: 286441-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR