(1H-Pyrazolo[3,4-b]pyridin-5-yl)boronic acid 95%

Supplier: Ambeed
Warning

A529513-1G A529513-250MG A529513-100MG
77156-992EA 351.23 USD
77156-992 77572-684 77684-372
(1H-Pyrazolo[3,4-b]pyridin-5-yl)boronic acid 95%
(1H-Pyrazolo[3,4-b]pyridin-5-yl)boronic acid

Precaución: For research use only.

Formula: C₆H₆BN₃O₂
MW: 162.94 g/mol
MDL Number: MFCD18250847
CAS Number: 1417985-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR