7-Aminopyrazolo[1,5-a]pyrimidine-6-carbonitrile 97%

Supplier: Ambeed
Danger

Synonyms: 7-Amino-6-cyanopyrazolo[1,5-a]pyrimidine

A528591-1G A528591-250MG A528591-5G
77156-982EA 77 USD
77156-982 77156-984 77901-728
7-Aminopyrazolo[1,5-a]pyrimidine-6-carbonitrile 97%
7-Aminopyrazolo[1,5-a]pyrimidine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₅
MDL Number: MFCD00104599
CAS Number: 89975-57-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR