Diatrizoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Di(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Bis(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Diacetamido-2,4,6-triiodobenzoic acid
A528364-1G
A528364-250MG
A528364-25G
A528364-5G
77156-976EA
7.21
USD
77156-976
77156-978
77156-980
77686-361
Diatrizoic acid 98%
Diatrizoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₉I₃N₂O₄ MW: 613.92 g/mol |
MDL Number:
MFCD00069960 CAS Number: 117-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...