1-Bromo-3,4,5-trifluorobenzene 98%

Supplier: Ambeed
Danger

A527209-10G A527209-5G A527209-25G A527209-100G
77156-968EA 6 USD
77156-968 77156-970 77551-590 77613-656
1-Bromo-3,4,5-trifluorobenzene 98%
1-Bromo-3,4,5-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₃
MDL Number: MFCD00070738
CAS Number: 138526-69-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR