C-(1-Benzylpyrrolidin-3-yl)methylamine 98%

Supplier: Ambeed
Danger

A525172-1G A525172-250MG A525172-25G A525172-5G
77156-950EA 57 USD
77156-950 77156-952 77156-954 77156-956
C-(1-Benzylpyrrolidin-3-yl)methylamine 98%

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂
MDL Number: MFCD06411056
CAS Number: 93138-61-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR