CH-223191 98%

Supplier: Ambeed
Warning

A525118-10MG A525118-5MG A525118-1MG
77156-946EA 90.01 USD
77156-946 77156-948 77613-652
CH-223191 98%
CH-223191

Precaución: for Research Use Only

Formula: C₁₉H₁₉N₅O
MDL Number: MFCD00377884
CAS Number: 301326-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR