2-mercaptobenzo[d]thiazol-6-ol 97%

Supplier: Ambeed
Warning

A524111-250MG A524111-5G
77156-934EA 29.87 USD
77156-934 77156-936
2-mercaptobenzo[d]thiazol-6-ol 97%
2-mercaptobenzo[d]thiazol-6-ol

Precaución: for Research Use Only

Formula: C₇H₅NOS₂
MW: 183.25 g/mol
MDL Number: MFCD23099217
CAS Number: 74537-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR