trans-4-(5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)cyclohexanol 98%

Supplier: Ambeed
Warning

A522576-100MG A522576-1G A522576-250MG A522576-5G
77156-920EA 12.36 USD
77156-920 77156-922 77156-924 77156-926
trans-4-(5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)cyclohexanol 98%
trans-4-(5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrClN₃O
MW: 330.60 g/mol
MDL Number: MFCD27987955
CAS Number: 1630906-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR