7-Oxa-2-azaspiro[4.5]decane oxalate(2:1) 95%

Supplier: Ambeed
Warning

A522077-100MG A522077-250MG
77156-916EA 131 USD
77156-916 77156-918
7-Oxa-2-azaspiro[4.5]decane oxalate(2:1) 95%
7-Oxa-2-azaspiro[4.5]decane oxalate(2:1)

Precaución: for Research Use Only

Formula: C₁₈H₃₂N₂O₆
MDL Number: MFCD28166296
CAS Number: 1630906-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR