6-Bromo-5-hydroxy-2-methyl-2H-indazole 98%
Supplier: Ambeed
|
Warning
|
A521508-100MG
A521508-1G
A521508-250MG
77156-910EA
46
USD
77156-910
77156-912
77156-914
6-Bromo-5-hydroxy-2-methyl-2H-indazole 98%
6-Bromo-5-hydroxy-2-methyl-2H-indazole
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂O |
MDL Number:
MFCD23106323 CAS Number: 1403767-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...