5-Bromo-1-(2-fluoro-6-(trifluoromethyl)benzyl)-6-methylpyrimidine-2,4(1H,3H)-dione 95%
Supplier: Ambeed
|
Warning
|
A521045-100G
A521045-1G
A521045-250MG
A521045-25G
A521045-5G
77156-878EA
302.82
USD
77156-878
77156-880
77156-882
77156-884
77156-886
5-Bromo-1-(2-fluoro-6-(trifluoromethyl)benzyl)-6-methylpyrimidine-2,4(1H,3H)-dione 95%
5-Bromo-1-(2-fluoro-6-(trifluoromethyl)benzyl)-6-methylpyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only
Formula:
C₁₃H₉BrF₄N₂O₂ MW: 381.12 g/mol |
MDL Number:
MFCD12546646 CAS Number: 830346-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...