(2,6-Dibromopyridin-4-yl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dibromo-4-hydroxymethylpyridine

A520804-1G A520804-250MG
77156-874EA 55.62 USD
77156-874 77156-876
(2,6-Dibromopyridin-4-yl)methanol 98%
(2,6-Dibromopyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MW: 266.91 g/mol
MDL Number: MFCD09836667
CAS Number: 223463-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR