6-Chlorochroman-4-amine 98%

Supplier: Ambeed
Warning

A518381-100MG A518381-250MG
77156-856EA 22 USD
77156-856 77156-858
6-Chlorochroman-4-amine 98%
6-Chlorochroman-4-amine

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MDL Number: MFCD07371530
CAS Number: 765880-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR