5-Bromo-1H-indazol-7-amine 95%

Supplier: Ambeed
Warning

A517996-100MG A517996-1G A517996-250MG A517996-5G
77156-848EA 30.9 USD
77156-848 77156-850 77156-852 77156-854
5-Bromo-1H-indazol-7-amine 95%
5-Bromo-1H-indazol-7-amine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.04 g/mol
MDL Number: MFCD13177026
CAS Number: 316810-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR