4-(Aminomethyl)oxazole Hydrochloride 97%

Supplier: Ambeed
Warning

A515919-100MG A515919-1G
77156-844EA 70.04 USD
77156-844 77156-846
4-(Aminomethyl)oxazole Hydrochloride 97%
4-(Aminomethyl)oxazole Hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₂O
MW: 134.56 g/mol
MDL Number: MFCD08437648
CAS Number: 847490-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR