trans-4-Cyanocyclohexylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (1r,4r)-4-aminocyclohexane-1-carbonitrile hydrochloride, trans-4-Aminocyclohexanecarbonitrile hydrochloride

A515765-1G A515765-250MG A515765-100MG A515765-5G
77156-840EA 335 USD
77156-840 77156-842 77686-344 77901-341
trans-4-Cyanocyclohexylamine hydrochloride 98%
trans-4-Cyanocyclohexylamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂N₂·ClH
MW: 160.645 g/mol
MDL Number: MFCD21643021
CAS Number: 873537-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR