(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S,S)-ip-FOXAP, (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine, (S,S)-[2-(4'-Isopropyloxazolin-2'yl)ferrocenyl]diphenylphosphine

A515747-1G A515747-5G
77156-836EA 67 USD
77156-836 77156-838
(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene ≥98%, ee 99%
(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene

Precaución: for Research Use Only

Formula: C₂₈H₂₈FeNOP
MW: 481.36 g/mol
MDL Number: MFCD15144869
CAS Number: 163169-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR