tert-Butyl-1H-pyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A515437-100MG A515437-250MG A515437-1G
77156-832EA 16 USD
77156-832 77156-834 77893-050
tert-Butyl-1H-pyrazole-4-carboxylate 95%
tert-Butyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD15976389
CAS Number: 611239-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR