5-Oxa-2,8-diazaspiro[3.5]nonane, N2-BOC protected 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-oxa-2,8-diazaspiro[3.5]nonane-2-carboxylate

A514533-100MG A514533-1G A514533-250MG
77156-808EA 130.81 USD
77156-808 77156-810 77156-812
5-Oxa-2,8-diazaspiro[3.5]nonane, N2-BOC protected 98%
5-Oxa-2,8-diazaspiro[3.5]nonane, N2-BOC protected

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₃
MW: 228.288 g/mol
MDL Number: MFCD14581202
CAS Number: 1251011-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR