4-(Aminomethyl)tetrahydro-2H-pyran-4-carbonitrile 95%

Supplier: Ambeed
Warning

A511538-1G A511538-250MG
77156-762EA 407.88 USD
77156-762 77686-332
4-(Aminomethyl)tetrahydro-2H-pyran-4-carbonitrile 95%
4-(Aminomethyl)tetrahydro-2H-pyran-4-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₁₂N₂O
MW: 140.18 g/mol
MDL Number: MFCD19215914
CAS Number: 1263374-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR