3-Ethylbenzophenone 95%

Supplier: Ambeed
Warning

A511261-1G A511261-250MG A511261-5G
77156-754EA 75 USD
77156-754 77156-756 77156-758
3-Ethylbenzophenone 95%
3-Ethylbenzophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O
MDL Number: MFCD01662328
CAS Number: 66067-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR