4-(2-Methyl-4-thiazolyl)phenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Methyl-4-thiazolyl)phenol [for Biochemical Research]

A510877-100MG A510877-1G A510877-250MG A510877-5G
77156-746EA 42.23 USD
77156-746 77156-748 77156-750 77156-752
4-(2-Methyl-4-thiazolyl)phenol 98%
4-(2-Methyl-4-thiazolyl)phenol

Precaución: for Research Use Only

Formula: C₁₀H₉NOS
MW: 191.25 g/mol
MDL Number: MFCD02642878
CAS Number: 30686-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR