(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-carbamoylphenyl)propanoic acid 97%
Supplier: Ambeed
|
Warning
|
A510559-100MG
A510559-1G
A510559-250MG
A510559-5G
77156-738EA
49.44
USD
77156-738
77156-740
77156-742
77156-744
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-carbamoylphenyl)propanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-carbamoylphenyl)propanoic acid
Precaución: for Research Use Only
Formula:
C₂₅H₂₂N₂O₅ MW: 430.45 g/mol |
MDL Number:
MFCD06659144 CAS Number: 959573-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...