(5-Methylthiophen-3-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A509547-100MG A509547-1G A509547-250MG A509547-5G
77156-718EA 35.02 USD
77156-718 77156-720 77156-722 77156-724
(5-Methylthiophen-3-yl)boronic acid ≥95%
(5-Methylthiophen-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₇BO₂S
MW: 141.98 g/mol
MDL Number: MFCD18250237
CAS Number: 930303-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR