2-Bromo-4-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A509013-100MG A509013-1G A509013-250MG A509013-5G
77156-708EA 25.75 USD
77156-708 77156-710 77156-712 77156-714
2-Bromo-4-hydroxybenzoic acid 95%
2-Bromo-4-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrO₃
MW: 217.017 g/mol
MDL Number: MFCD16999959
CAS Number: 28547-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR