3-Chloro-4-fluorobenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,2-Dioxaborolane, 2-(3-chloro-4-fluorophenyl)-4,4,5,5-tetramethyl-, 3-Chloro-4-fluorophenylboronic acid pinacol ester, 2-(3-Chloro-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A508173-25G A508173-5G
77156-704EA 72.1 USD
77156-704 77156-706
3-Chloro-4-fluorobenzeneboronic acid pinacol ester 98%
3-Chloro-4-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClFO₂
MW: 256.51 g/mol
MDL Number: MFCD11111387
CAS Number: 635305-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR