4'-Amino-2'-chloroacetophenone 98+%

Supplier: Ambeed
Warning

A507243-1G A507243-250MG A507243-5G
77156-698EA 52.53 USD
77156-698 77156-700 77156-702
4'-Amino-2'-chloroacetophenone 98+%
4'-Amino-2'-chloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.608 g/mol
MDL Number: MFCD11848240
CAS Number: 72531-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR