(S)-2-((tert-Butoxycarbonyl)amino)-2-cycloheptylacetic acid 98%

Supplier: Ambeed
Warning

A506262-100MG A506262-1G A506262-250MG
77156-676EA 50.47 USD
77156-676 77156-678 77156-680
(S)-2-((tert-Butoxycarbonyl)amino)-2-cycloheptylacetic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cycloheptylacetic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₄
MW: 271.35 g/mol
MDL Number: MFCD16251538
CAS Number: 1228542-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR