Ethyl 4-fluoro-2-methylbenzoate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-methylbenzoic acid ethyl ester

A505117-100G A505117-25G A505117-5G
77156-668EA 178.19 USD
77156-668 77156-670 77156-672
Ethyl 4-fluoro-2-methylbenzoate 95%
Ethyl 4-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MW: 182.19 g/mol
MDL Number: MFCD06204529
CAS Number: 167758-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR