4,4'-Dibromo-4''-phenyltriphenylamine 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Biphenylyl)-N,N-bis(4-bromophenyl)amine

A504865-1G A504865-250MG A504865-5G
77156-660EA 16 USD
77156-660 77156-662 77156-664
4,4'-Dibromo-4''-phenyltriphenylamine 98%
4,4'-Dibromo-4''-phenyltriphenylamine

Precaución: for Research Use Only

Formula: C₂₄H₁₇Br₂N
MW: 479.22 g/mol
MDL Number: MFCD08276360
CAS Number: 884530-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR